accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClCH3NNNSFFF

CAS RN 440-17-5

IUPAC Name: dihydrogen 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)-10H-phenothiazine dichloride
Synonyms: trifluoperazine hydrochloride trifluoroperazine dihydrochloride trifluoperazine dihydrochloride trifluoperazine hcl trifluperazine dihydrochloride
Molecular Weight (g/mol): 480.42
Molecular Formula: C21H26Cl2F3N3S
InChi Key: BXDAOUXDMHXPDI-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CN1CCN(CCCN2C3=CC=CC=C3SC3=CC=C(C=C23)C(F)(F)F)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results