accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO

CAS RN 4401-20-1

IUPAC Name: 2-cycloheptylacetic acid
Synonyms: cycloheptylacetic acid
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: DQNWKASPZFJVMJ-UHFFFAOYSA-N
SMILES: OC(=O)CC1CCCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results