accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4402-24-8

IUPAC Name: diethyl (3-aminopropyl)phosphonate
Synonyms: diethyl 3-aminopropylphosphonate
Molecular Weight (g/mol): 195.2
Molecular Formula: C7H18NO3P
InChi Key: DQORFBNFNLHVIM-UHFFFAOYSA-N
SMILES: CCOP(=O)(CCCN)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results