accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4408-78-0

IUPAC Name: 2-phosphonoacetic acid
Synonyms: phosphonoacetic acid
Molecular Weight (g/mol): 140.03
Molecular Formula: C2H5O5P
InChi Key: XUYJLQHKOGNDPB-UHFFFAOYSA-N
SMILES: OC(=O)CP(O)(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)