accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(E)

CAS RN 4411-89-6

IUPAC Name: (2E)-2-phenylbut-2-enal
Synonyms: (2E)-2-phenylbut-2-enal
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: DYAOGZLLMZQVHY-MBXJOHMKSA-N
SMILES: C\C=C(\C=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results