accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4424-06-0

IUPAC Name: 3,10,17,24-tetraazaoctacyclo[13.13.2.0²,¹⁰.0⁴,⁹.0¹²,²⁹.0¹⁶,²⁴.0¹⁸,²³.0²⁶,³⁰]triaconta-1(28),2,4,6,8,12,14,16,18,20,22,26,29-tridecaene-11,25-dione
Synonyms: perinone
Molecular Weight (g/mol): 412.41
Molecular Formula: C26H12N4O2
InChi Key: DGBWPZSGHAXYGK-UHFFFAOYSA-N
SMILES: O=C1N2C3=CC=CC=C3N=C2C2=CC=C3C(=O)N4C5=CC=CC=C5N=C4C4=CC=C1C2=C34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results