accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH3CFFFK

CAS RN 44248-07-9

IUPAC Name: potassium ethyltrifluoroboranuide
Synonyms: potassium ethyltrifluoroboranuide
Molecular Weight (g/mol): 135.97
Molecular Formula: C2H5BF3K
InChi Key: GIIPADVFWNGSKY-UHFFFAOYSA-N
SMILES: [K+].CC[B-](F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results