accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNFCl(E)

CAS RN 443-33-4

IUPAC Name: (E)-N-[(2-chloro-6-fluorophenyl)methylidene]hydroxylamine
Synonyms: (E)-N-[(2-chloro-6-fluorophenyl)methylidene]hydroxylamine
Molecular Weight (g/mol): 173.57
Molecular Formula: C7H5ClFNO
InChi Key: OBJHLLOVMKKXDI-ONNFQVAWSA-N
SMILES: O\N=C\C1=C(F)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results