accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CNHOHOOSOOO

CAS RN 4430-18-6

IUPAC Name: sodium 2-[(4-hydroxy-9,10-dioxo-9,10-dihydroanthracen-1-yl)amino]-5-methylbenzene-1-sulfonate
Synonyms: acid violet 43
Molecular Weight (g/mol): 431.39
Molecular Formula: C21H14NNaO6S
InChi Key: GTKIEPUIFBBXJQ-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC=C(NC2=CC=C(O)C3=C2C(=O)C2=CC=CC=C2C3=O)C(=C1)S([O-])(=O)=O
Color 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results