accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4430-47-1

IUPAC Name: 5-(isothiocyanatomethyl)-2H-1,3-benzodioxole
Synonyms: 5-(isothiocyanatomethyl)-2H-1,3-benzodioxole
Molecular Weight (g/mol): 193.22
Molecular Formula: C9H7NO2S
InChi Key: PUJWRDBPAFJUJW-UHFFFAOYSA-N
SMILES: S=C=NCC1=CC=C2OCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results