accessibility menu, dialog, popup

UserName

Viewing profile as user:
SCNNCS

CAS RN 4430-51-7

IUPAC Name: 1,4-diisothiocyanatobutane
Synonyms: 1,4-diisothiocyanatobutane
Molecular Weight (g/mol): 172.26
Molecular Formula: C6H8N2S2
InChi Key: RRSISCMPUAGVJN-UHFFFAOYSA-N
SMILES: S=C=NCCCCN=C=S
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)