accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4430-75-5

IUPAC Name: octahydro-1H-pyrido[1,2-a]pyrazine
Synonyms: octahydro-1H-pyrido[1,2-a]pyrazine
Molecular Weight (g/mol): 140.23
Molecular Formula: C8H16N2
InChi Key: ONHPOXROAPYCGT-UHFFFAOYNA-N
SMILES: C1CCN2CCNCC2C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)