accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4431-00-9

IUPAC Name: triammonium 5-[(3-carboxylato-4-hydroxyphenyl)(3-carboxylato-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoate
Synonyms: aurintricarboxylic acid
Molecular Weight (g/mol): 475.45
Molecular Formula: C22H25N3O9
InChi Key: UDPQMNAHKKTHHI-UHFFFAOYNA-N
SMILES: [NH4+].[NH4+].[NH4+].OC1=CC=C(C=C1C([O-])=O)C(C1C=CC(=O)C(=C1)C([O-])=O)C1=CC=C(O)C(=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results