CAS RN 4437-01-8
CAS RN 4437-01-8
IUPAC Name:
2,5,8,11,14,17,20-heptaoxadocosan-22-ol
Synonyms:
2,5,8,11,14,17,20-heptaoxadocosan-22-ol
Molecular Weight (g/mol):
340.41
Molecular Formula:
C15H32O8
InChi Key:
AGWKUHGLWHMYTG-UHFFFAOYSA-N
SMILES:
COCCOCCOCCOCCOCCOCCOCCO
1
–
1
of
1
results
1
Heptaethylene Glycol Monomethyl Ether 97.0+%, TCI America™
CAS: 4437-01-8 Molecular Formula: C15H32O8 Molecular Weight (g/mol): 340.41 MDL Number: MFCD06201006 InChI Key: AGWKUHGLWHMYTG-UHFFFAOYSA-N Synonym: mPEG7-Alcohol PubChem CID: 526555 IUPAC Name: 2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol SMILES: COCCOCCOCCOCCOCCOCCOCCO