accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 443882-99-3

IUPAC Name: 2-chloro-1-[(3-fluorophenyl)methoxy]-4-nitrobenzene
Synonyms: 2-chloro-1-[(3-fluorophenyl)methoxy]-4-nitrobenzene
Molecular Weight (g/mol): 281.67
Molecular Formula: C13H9ClFNO3
InChi Key: SFTHVDYRPHJAND-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(OCC2=CC(F)=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results