accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4439-02-5

IUPAC Name: 2-(2H-1,3-benzodioxol-5-yl)acetonitrile
Synonyms: 2-(2H-1,3-benzodioxol-5-yl)acetonitrile
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: ZQPBOYASBNAXOZ-UHFFFAOYSA-N
SMILES: N#CCC1=CC=C2OCOC2=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)