accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4441-30-9

IUPAC Name: 3-(morpholin-4-yl)propan-1-ol
Synonyms: 3-(morpholin-4-yl)propan-1-ol
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: VZKSLWJLGAGPIU-UHFFFAOYSA-N
SMILES: OCCCN1CCOCC1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)