accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOH

CAS RN 4441-56-9

IUPAC Name: cyclohexylboronic acid
Synonyms: cyclohexylboronic acid
Molecular Weight (g/mol): 127.98
Molecular Formula: C6H13BO2
InChi Key: XDRVAZAFNWDVOE-UHFFFAOYSA-N
SMILES: OB(O)C1CCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)