accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4442-54-0

IUPAC Name: 2,3-dihydro-1,4-benzodioxine-6-carboxylate
Synonyms: 2,3-dihydro-1,4-benzodioxine-6-carboxylate
Molecular Weight (g/mol): 179.15
Molecular Formula: C9H7O4
InChi Key: JWZQJTGQFHIRFQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C2OCCOC2=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)