CAS RN 444722-95-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Cdk1/2 Inhibitor II, NU6102
The Cdk1/2 Inhibitor II, NU6102, also referenced under CAS 444722-95-6, controls the biological activity of Cdk1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.