accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNNNCH3NNH3CH3CSOONH2

CAS RN 444731-52-6

IUPAC Name: 5-({4-[(2,3-dimethyl-2H-indazol-6-yl)(methyl)amino]pyrimidin-2-yl}amino)-2-methylbenzene-1-sulfonamide
Synonyms: pazopanib
Molecular Weight (g/mol): 437.52
Molecular Formula: C21H23N7O2S
InChi Key: CUIHSIWYWATEQL-UHFFFAOYSA-N
SMILES: CN(C1=CC2=NN(C)C(C)=C2C=C1)C1=NC(NC2=CC(=C(C)C=C2)S(N)(=O)=O)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results