accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 445303-67-3

BOOH3CCH3H3CH3CFFFF

CAS RN 445303-67-3

IUPAC Name: 2-[3-fluoro-4-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[3-fluoro-4-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 290.06
Molecular Formula: C13H15BF4O2
InChi Key: FPCSARGPMNDYAO-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(F)=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results