accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSCH3

CAS RN 4455-13-4

IUPAC Name: ethyl 2-(methylsulfanyl)acetate
Synonyms: ethyl 2-(methylsulfanyl)acetate
Molecular Weight (g/mol): 134.19
Molecular Formula: C5H10O2S
InChi Key: MDIAKIHKBBNYHF-UHFFFAOYSA-N
SMILES: CCOC(=O)CSC
Boiling Point 
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)