accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrBr

CAS RN 4457-72-1

IUPAC Name: 1,5-dibromo-3-methylpentane
Synonyms: 1,5-dibromo-3-methylpentane
Molecular Weight (g/mol): 243.97
Molecular Formula: C6H12Br2
InChi Key: YDPZWUMQKMLLHC-UHFFFAOYSA-N
SMILES: CC(CCBr)CCBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results