accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 446-10-6

IUPAC Name: 4-fluoro-1-methyl-2-nitrobenzene
Synonyms: 2-nitro-4-fluorotoluene
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: SKWTUNAAJNDEIK-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results