CAS RN 446-24-2
IUPAC Name:
2-fluorobenzohydrazide
Synonyms:
2-fluorobenzohydrazide
Molecular Weight (g/mol):
154.14
Molecular Formula:
C7H7FN2O
InChi Key:
YJCCKQQVXNNAAR-UHFFFAOYSA-N
SMILES:
NNC(=O)C1=CC=CC=C1F