accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOFF

CAS RN 446-35-5

IUPAC Name: 2,4-difluoro-1-nitrobenzene
Synonyms: benzene, 2,4-difluoro-1-nitro-
Molecular Weight (g/mol): 159.09
Molecular Formula: C6H3F2NO2
InChi Key: RJXOVESYJFXCGI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C=C1F
Boiling Point 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)