accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 446-86-6

IUPAC Name: 6-[(1-methyl-4-nitro-1H-imidazol-5-yl)sulfanyl]-7H-purine
Synonyms: azathioprine ccucol azothioprine azathioprine sodium muran azatioprin imurek imurel imuran azathioprin
Molecular Weight (g/mol): 277.26
Molecular Formula: C9H7N7O2S
InChi Key: LMEKQMALGUDUQG-UHFFFAOYSA-N
SMILES: CN1C=NC(=C1SC1=C2NC=NC2=NC=N1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (5)
Grade 
  • (1)
  • (5)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (3)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (5)
  • (5)