accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNOO

CAS RN 446044-44-6

IUPAC Name: 2-{1,3-dioxo-1H,2H,3H-naphtho[2,3-f]isoindol-2-yl}cyclohexane-1-carboxylic acid
Synonyms: 2-{1,3-dioxonaphtho[2,3-f]isoindol-2-yl}cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 373.41
Molecular Formula: C23H19NO4
InChi Key: IIJKRPVAVXANME-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCCC1N1C(=O)C2=C(C=C3C=C4C=CC=CC4=CC3=C2)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)