accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 446242-37-1

IUPAC Name: N-(naphthalen-1-yl)-[1,1'-biphenyl]-4-amine
Synonyms: N-(naphthalen-1-yl)-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 295.39
Molecular Formula: C22H17N
InChi Key: KDADHQHDRSAQDY-UHFFFAOYSA-N
SMILES: N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results