CAS RN 4464-18-0
IUPAC Name:
[3,5-bis(hydroxymethyl)phenyl]methanol
Synonyms:
[3,5-bis(hydroxymethyl)phenyl]methanol
Molecular Weight (g/mol):
168.19
Molecular Formula:
C9H12O3
InChi Key:
SQAMZFDWYRVIMG-UHFFFAOYSA-N
SMILES:
OCC1=CC(CO)=CC(CO)=C1
1
–
1
of
1
results
1
1,3,5-Benzenetrimethanol 95.0+%, TCI America™
CAS: 4464-18-0 Molecular Formula: C9H12O3 Molecular Weight (g/mol): 168.192 MDL Number: MFCD01108484 InChI Key: SQAMZFDWYRVIMG-UHFFFAOYSA-N Synonym: 1,3,5-Tris(hydroxymethyl)benzene PubChem CID: 2748048 IUPAC Name: [3,5-bis(hydroxymethyl)phenyl]methanol SMILES: C1=C(C=C(C=C1CO)CO)CO