accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3N

CAS RN 4467-06-5

IUPAC Name: 2-(4-methylphenyl)pyridine
Synonyms: 2-(4-methylphenyl)pyridine
Molecular Weight (g/mol): 169.23
Molecular Formula: C12H11N
InChi Key: KJNZQKYSNAQLEO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC=CC=N1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)