accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFN

CAS RN 447-60-9

IUPAC Name: 2-(trifluoromethyl)benzonitrile
Synonyms: A,A,A-trifluor-O-tolunitril
Molecular Weight (g/mol): 171.12
Molecular Formula: C8H4F3N
InChi Key: SOZGHDCEWOLLHV-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1C#N
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)