accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 447-61-0

IUPAC Name: 2-(trifluoromethyl)benzaldehyde
Synonyms: 2-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 174.12
Molecular Formula: C8H5F3O
InChi Key: ZDVRPKUWYQVVDX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1C=O
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (5)
Quantity 
  • (3)
  • (4)
  • (1)
Boiling Point 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (5)
  • (4)
  • (4)
  • (1)
  • (2)
  • (5)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)