accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHONOHO

CAS RN 4479-74-7

IUPAC Name: [2,2'-bipyridine]-6,6'-dicarboxylic acid
Synonyms: [2,2'-bipyridine]-6,6'-dicarboxylic acid
Molecular Weight (g/mol): 244.21
Molecular Formula: C12H8N2O4
InChi Key: DASAXWLMIWDYLQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=N1)C1=NC(=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results