accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 448-61-3

IUPAC Name: 2,4,6-triphenyl-1λ⁴-pyran-1-ylium; tetrafluoroboranuide
Synonyms: 2,4,6-triphenyl-1λ⁴-pyran-1-ylium tetrafluoroborate
Molecular Weight (g/mol): 396.19
Molecular Formula: C23H17BF4O
InChi Key: VQYPWMWEJGDSTF-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.C1=CC=C(C=C1)C1=CC(=[O+]C(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)