accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4482-01-3

IUPAC Name: 2H-1λ⁶,3λ⁶,2-benzodithiazole-1,1,3,3-tetrone
Synonyms: 2H-1λ⁶,3λ⁶,2-benzodithiazole-1,1,3,3-tetrone
Molecular Weight (g/mol): 219.23
Molecular Formula: C6H5NO4S2
InChi Key: QRWQFOHBHHIZKG-UHFFFAOYSA-N
SMILES: O=S1(=O)NS(=O)(=O)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)