accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOBrN

CAS RN 4487-59-6

IUPAC Name: 2-bromo-5-nitropyridine
Synonyms: 2-bromo-5-nitropyridine
Molecular Weight (g/mol): 203
Molecular Formula: C5H3BrN2O2
InChi Key: HUUFTVUBFFESEN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Br)N=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)