accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOO

CAS RN 4488-40-8

IUPAC Name: 4-methylnaphthalene-1-carboxylic acid
Synonyms: 4-methylnaphthalene-1-carboxylic acid
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: SIVYRLBDAPKADZ-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=C(C=C1)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)