accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOH

CAS RN 448967-90-6

IUPAC Name: (1-methyl-1H-indol-5-yl)methanol
Synonyms: (1-methylindol-5-yl)methanol
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: CEBLTQGXYITWTM-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC(CO)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results