CAS RN 4498-39-9
IUPAC Name:
4-methyl-2,3-dihydro-1,3-thiazole-2-thione
Synonyms:
4-methyl-3H-1,3-thiazole-2-thione
Molecular Weight (g/mol):
131.21
Molecular Formula:
C4H5NS2
InChi Key:
NLHAIPFBNQZTMY-UHFFFAOYSA-N
SMILES:
CC1=CSC(=S)N1
1
–
1
of
1
results
1
2-Mercapto-4-methylthiazole, 97%, Thermo Scientific™
CAS: 4498-39-9 Molecular Formula: C4H5NS2 Molecular Weight (g/mol): 131.21 MDL Number: MFCD00798571 InChI Key: NLHAIPFBNQZTMY-UHFFFAOYSA-N Synonym: 4-methylthiazole-2-thiol,4-methyl-1,3-thiazole-2-thiol,2-mercapto-4-methylthiazole,2 3h-thiazolethione, 4-methyl,4-methyl-2 3h-thiazolethione,4-methylthiazole-2 3h-thione,4-methyl-2-thiazolethiol,4-methyl-2-mercaptothiazole,4-methylthiazoline-2-thione,4-methyl-1,3-thiazole-2 3h-thione PubChem CID: 1201418 IUPAC Name: 4-methyl-2,3-dihydro-1,3-thiazole-2-thione SMILES: CC1=CSC(=S)N1