accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4498-67-3

IUPAC Name: 1H-indazole-3-carboxylic acid
Synonyms: 1H-indazole-3-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: BHXVYTQDWMQVBI-UHFFFAOYSA-N
SMILES: OC(=O)C1=NNC2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)