accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 450-89-5

IUPAC Name: 1-chloro-4-fluoro-2-methoxybenzene
Synonyms: 1-chloro-4-fluoro-2-methoxybenzene
Molecular Weight (g/mol): 160.57
Molecular Formula: C7H6ClFO
InChi Key: ULVPJKKERHMKLS-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results