accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4502-10-7

H3COHOH2N

CAS RN 4502-10-7

IUPAC Name: 1-(2-amino-3-hydroxyphenyl)ethan-1-one
Synonyms: 1-(2-amino-3-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: DIIASMSSGMRMQF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(O)=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results