accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 450368-20-4

IUPAC Name: 6-(pyrrolidin-1-yl)pyridine-2-carboxylic acid
Synonyms: 6-(pyrrolidin-1-yl)pyridine-2-carboxylic acid
Molecular Weight (g/mol): 192.22
Molecular Formula: C10H12N2O2
InChi Key: HXPHLYQBFPDHNK-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC(=CC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results