accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 451-46-7

IUPAC Name: ethyl 4-fluorobenzoate
Synonyms: ethyl 4-fluorobenzoate
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: UMPRJGKLMUDRHL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(F)C=C1
Boiling Point 
  • (5)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)