accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOF(E)

CAS RN 451-69-4

IUPAC Name: (2E)-3-(2-fluorophenyl)prop-2-enoic acid
Synonyms: cinnamic acid, o-fluoro-
Molecular Weight (g/mol): 166.15
Molecular Formula: C9H7FO2
InChi Key: IOUDZAFBPDDAMK-AATRIKPKSA-N
SMILES: OC(=O)\C=C\C1=CC=CC=C1F
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)