accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHONa

CAS RN 4510-76-3

IUPAC Name: sodium 2-oxo-1,2-dihydroquinolin-4-olate
Synonyms: sodium 2-oxo-1H-quinolin-4-olate
Molecular Weight (g/mol): 183.14
Molecular Formula: C9H6NNaO2
InChi Key: MWXMDMIYCCSYNP-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=CC(=O)NC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results