accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2COOFFFFF

CAS RN 45115-53-5

IUPAC Name: 2,2,3,3,3-pentafluoropropyl 2-methylprop-2-enoate
Synonyms: 2,2,3,3,3-pentafluoropropyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 218.12
Molecular Formula: C7H7F5O2
InChi Key: CLISWDZSTWQFNX-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC(F)(F)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)