accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 45120-30-7

IUPAC Name: 4-amino-5-(tert-butoxy)-5-oxopentanoic acid
Synonyms: 4-amino-5-(tert-butoxy)-5-oxopentanoic acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: QVAQMUAKTNUNLN-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C(N)CCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)